missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Mastercycler™ nexus GX2e, 230 V/50 – 60 Hz (GB)

Product Code. 15785675 Shop All Eppendorf Products
This item is not returnable. View return policy
Product Code. 15785675 Supplier Eppendorf™ Supplier No. 6338000047
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

If you’ve got a high PCR workload, turn to the Eppendorf Mastercycler nexus X2. With two blocks (32-well and 64-well), you can run two independent protocols at the same time using a variety of consumables up to 0.5 mL tubes. For higher throughput, network with up to 3 additional family members.

Mastercycler™ nexus GX2e, 230 V/50 – 60 Hz (GB)- Asymmetrically divided block for running different assays in parallel- Universal block for strips, 0.1 mL, 0.2 mL and 0.5 mL PCR tubes- Small footprint- With 8 column gradient for PCR optimization- E-mail notification- flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
TRUSTED_SUSTAINABILITY

Specifications

Thermal Accuracy ±0.2°C
Thermal Uniformity ≤±0.3°C
Dimensions 41.2 x 25 x 33 cm
Capacity 64/32 x 0.2 mL PCR Tubes or up to 45/19 x 0.5 mL PCR Tubes
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz
Product Type PCR
Thermal Range 4°C to 99°C
For Use With (Application) Standard PCR, Optimizing a PCR
Frequency 50 to 60 Hz
User Interface Intuitive
Wattage 700 W
Max. Ramp Rate 3°C/s (Measured at the Block)
No. of Programs >700
Heated Lid Yes
Weight 10.5 kg
Block Configurations 0.2/0.5 mL Tubes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.