missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Master Tips™ Micropipette Tips
GREENER_CHOICE

Product Code. 10777021 Shop All Eppendorf Products
Change view
Click to view available options
Master Tips™ Micropipette Tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10777021 Master Tips™ Micropipette Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10777021 Supplier Eppendorf™ Supplier No. 0030001320

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use these pipette tips for contamination free pipetting. Master Tips™ Micropipette Tips are ideal for accurate pipetting of liquids with high-vapor pressure.

  • Piston integrated in tip and ready for use
  • Racked, autoclavable at 121°C, 20 min.
  • For use with Biomaster™
TRUSTED_SUSTAINABILITY

Specifications

Description Syringe style, positive displacement tip with integrated piston
Volume (Metric) 20 μL
Color Gray
Autoclavable Autoclavable
Length (Metric) 52 mm
Certifications/Compliance Eppendorf Quality™
For Use With (Application) Accurate pipetting of viscous solutions or liquids with high vapour pressure
Features Integrated piston
Format Rack, 5 x 96
For Use With (Equipment) BioMaster
Product Line Master Tip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.