Learn More
MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 Mouse Monoclonal Antibody, Epredia™
Epredia™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.
£114.24 - £580.29
Specifications
| Antigen | MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 |
|---|---|
| Clone | M2-7C10 + M2-9E3 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Form | Concentration | Price | Quantity & Availability | |||
|
12642127
|
Epredia
MS-716-P0 |
100 μL | Concentrated | 200μg/mL | |||||
|
12662127
|
Epredia
MS-716-P1ABX |
100 μL | Concentrated | 1mg/mL | |||||
|
12672127
|
Epredia
MS-716-PABX |
200 μL | Concentrated | 1mg/mL | |||||
|
12652127
|
Epredia
MS-716-P1 |
500 μL | Concentrated | 200μg/mL |
£368.00 £346.20 / 500µL Save £21.80 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12632127
|
Epredia
MS-716-P |
1 mL | Concentrated | 200μg/mL |
£616.00 £580.29 / 1mL Save £35.71 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
12612137
|
Epredia
MS-716-R7 |
7 mL | Prediluted | ||||||
Description
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.Host Species: Mouse
Clone: M2-7C10 + M2-9E3
Isotype: IgG2b/κ + IgG2b/κ
Species Reactivity: Human and Horse. Others not known.
Epitope: Not determined
Immunogen: Recombinant hMART-1 protein
Molecular Weight: 18kDa
Positive Control: CAC1 melanoma cells and Melanomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
| MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RUO | |
| Recombinant hMART-1 protein |
| M2-7C10 + M2-9E3 | |
| Monoclonal | |
| Mouse | |
| Cancer and Tumor Biology | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.