Learn More
MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 Mouse Monoclonal Antibody, Epredia™
Epredia™ MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-716-P1ABX
Description
MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-Ais a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum.Host Species: Mouse
Clone: M2-7C10 + M2-9E3
Isotype: IgG2b/κ + IgG2b/κ
Species Reactivity: Human and Horse. Others not known.
Epitope: Not determined
Immunogen: Recombinant hMART-1 protein
Molecular Weight: 18kDa
Positive Control: CAC1 melanoma cells and Melanomas
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
| MART-1/Melan-A (Melanoma Marker) (Cocktail) Ab-3 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| 100 μL | |
| Cancer and Tumor Biology | |
| Equine, Human | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| M2-7C10 + M2-9E3 | |
| Unconjugated | |
| Recombinant hMART-1 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.