missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | MARCH9 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
44264 Polyclonal specifically detects 44264 in Human samples. It is validated for Western Blot.Specifications
| MARCH9 | |
| Polyclonal | |
| Rabbit | |
| Zinc Finger | |
| E3 ubiquitin-protein ligase MARCH9, EC 6.3.2.-, FLJ36578, MARCH-IX, membrane-associated ring finger (C3HC4) 9, Membrane-associated RING finger protein 9, Membrane-associated RING-CH protein IX, RNF179RING finger protein 179 | |
| Synthetic peptides corresponding to MARCH9(membrane-associated ring finger (C3HC4) 9) The peptide sequence was selected from the middle region of 40246 (NP_612405). Peptide sequence SRTGPTSGATSRPPAAQRMRTLLPQRCGYTILHLLGQLRPPDARSSSHSG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q86YJ5 | |
| 92979 | |
| IgG | |
| 38 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title