missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCH9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59692
This item is not returnable.
View return policy
Description
44264 Polyclonal specifically detects 44264 in Human samples. It is validated for Western Blot.
Specifications
| MARCH9 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| E3 ubiquitin-protein ligase MARCH9, EC 6.3.2.-, FLJ36578, MARCH-IX, membrane-associated ring finger (C3HC4) 9, Membrane-associated RING finger protein 9, Membrane-associated RING-CH protein IX, RNF179RING finger protein 179 | |
| Rabbit | |
| 38 kDa | |
| 100 μL | |
| Zinc Finger | |
| 92979 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q86YJ5 | |
| MARCHF9 | |
| Synthetic peptides corresponding to MARCH9(membrane-associated ring finger (C3HC4) 9) The peptide sequence was selected from the middle region of 40246 (NP_612405). Peptide sequence SRTGPTSGATSRPPAAQRMRTLLPQRCGYTILHLLGQLRPPDARSSSHSG. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody prior to reconstitution. Add 50μL of distilled water to a final antibody concentration of 1mg/mL. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction