missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP38 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45571
This item is not returnable.
View return policy
Description
FKBP38 Polyclonal specifically detects FKBP38 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FKBP38 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 0.5% BSA, 5 mM EDTA, 30% Glycerol with 0.03% Proclin 300 | |
| 38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 0.5 - 4 ug/mL | |
| AY225340 | |
| FKBP8 | |
| Synthetic peptide surrounding amino acid 344 of mouse FKBP38. | |
| 0.1 mg | |
| Signal Transduction | |
| 23770 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction