missing translation for 'onlineSavingsMsg'
Learn More

FKBP38 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£139.00 - £375.80

Specifications

Antigen FKBP38
Concentration 0.5 mg/ml
Dilution Western Blot 0.5 - 4 ug/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18232383
View Documents
Novus Biologicals
NBP1-45571-0.025MG
0.025 mg
£139.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18207212
View Documents
Novus Biologicals
NBP1-45571
0.1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

FKBP38 Polyclonal specifically detects FKBP38 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

FKBP38
Western Blot 0.5 - 4 ug/mL
Polyclonal
Rabbit
Signal Transduction
AY225340
23770
Synthetic peptide surrounding amino acid 344 of mouse FKBP38.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/ml
Western Blot
Unconjugated
RUO
Human, Mouse, Rat
38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase
FKBP8
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.