missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK2 Antibody (7H4.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£148.00 - £501.00
Specifications
| Antigen | MAPKAPK2 |
|---|---|
| Clone | 7H4.2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:20 |
| Applications | Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18125984
|
Novus Biologicals
NBP2-29767-0.05MG |
0.05 mg |
£148.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18111402
|
Novus Biologicals
NBP2-29767 |
0.2 mg |
£501.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAPKAPK2 Monoclonal specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| MAPKAPK2 | |
| 1 mg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat, Mouse | |
| EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 | |
| MAPKAPK2 | |
| IgG | |
| Affinity Purified | |
| 45 kDa |
| 7H4.2 | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:20 | |
| Monoclonal | |
| Mouse | |
| Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| P49137 | |
| 9261 | |
| KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title