missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK2 Antibody (7H4.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29767-0.05MG
This item is not returnable.
View return policy
Description
MAPKAPK2 Monoclonal specifically detects MAPKAPK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| MAPKAPK2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:20 | |
| P49137 | |
| MAPKAPK2 | |
| KLH-conjugated synthetic peptide of the C-terminus of the long form of human MAPKAP-K2. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat, Mouse | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 7H4.2 | |
| Unconjugated | |
| 0.2 mg antibody with PBS with 0.1% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, MAP kinase-activated protein kinase 2, MAPK-activated protein kinase 2, MAPKAP kinase 2, MAPKAPK-2, mitogen-activated protein kinase-activated protein kinase 2, MK2 | |
| Mouse | |
| 45 kDa | |
| 0.05 mg | |
| Breast Cancer, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction | |
| 9261 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction