missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAPK8 (JNK1), His Tag Recombinant Protein
Human MAPK8 (JNK1), His Tag Recombinant Protein
Brand: Invitrogen™ PV3319
Code : 50
Additional Details : Weight : 3.36000kg
Description
MAPK8 (JNK1) is activated by the presence of malfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity.Specifications
NP_002741.1 | |
5599 | |
10μg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: CMGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
mitogen-activated protein kinase 8 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
47.6 kDa | |
MAPK8 (JNK1) | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
MAPK8 | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.