missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAPK8 (JNK1), His Tag Recombinant Protein
Human MAPK8 (JNK1), His Tag Recombinant Protein
£437.00 - £6900.00
Specifications
Accession Number | NP_002741.1 |
---|---|
Gene Alias | MAPK8 (JNK1) |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11583427
|
Invitrogen™
PV3319 |
10μg |
£437.00
10µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10053082
|
Invitrogen™
PV3593 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418453
|
Invitrogen™
PR5340A |
100 μg |
£1600.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAPK8 (JNK1) is activated by the presence of malfolded proteins in the endoplasmic reticulum. It is also a crucial mediator of insulin resistance and obesity.Specifications
NP_002741.1 | |
Human | |
Baculovirus |
MAPK8 (JNK1) | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.