missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | MAPK15 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MAPK15 Polyclonal specifically detects MAPK15 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MAPK15 | |
| Polyclonal | |
| Rabbit | |
| Q8TD08 | |
| 225689 | |
| Synthetic peptides corresponding to MAPK15(mitogen-activated protein kinase 15) The peptide sequence was selected from the middle region of MAPK15. Peptide sequence GHDPAEHESPRAAKNVPRQNSAPLLQTALLGNGERPPGAKEAPPLTLSLV. | |
| Primary |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| EC 2.7.11, ERK-7, ERK7MAPK 15, ERK-8, ERK8EC 2.7.11.24, extracellular regulated kinase 8 delta, extracellular signal regulated kinase 8, Extracellular signal-regulated kinase 7, Extracellular signal-regulated kinase 8, MAP kinase 15, mitogen-activated protein kinase 15 | |
| MAPK15 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title