missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56789
This item is not returnable.
View return policy
Description
MAPK15 Polyclonal specifically detects MAPK15 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MAPK15 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.7.11, ERK-7, ERK7MAPK 15, ERK-8, ERK8EC 2.7.11.24, extracellular regulated kinase 8 delta, extracellular signal regulated kinase 8, Extracellular signal-regulated kinase 7, Extracellular signal-regulated kinase 8, MAP kinase 15, mitogen-activated protein kinase 15 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 225689 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:200 | |
| Q8TD08 | |
| MAPK15 | |
| Synthetic peptides corresponding to MAPK15(mitogen-activated protein kinase 15) The peptide sequence was selected from the middle region of MAPK15. Peptide sequence GHDPAEHESPRAAKNVPRQNSAPLLQTALLGNGERPPGAKEAPPLTLSLV. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%;. | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction