missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAPK10 (JNK3) Recombinant Human Protein
MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs).
£1602.00 - £6900.00
Specifications
Accession Number | NP_002744 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 76.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488493
|
Invitrogen™
PR6983B |
100μg |
£1602.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448733
|
Invitrogen™
PV4564 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4.
Specifications
NP_002744 | |
76.6kDa | |
RUO - research use only | |
MAPK10 (JNK3) | |
Human | |
GST Tag | |
Frozen |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAP Kinase Family | |
Recombinant | |
Baculovirus |