missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAPK10 (JNK3) Recombinant Human Protein

MAPK10 (JNK3) is a neuronal-specific form of c-Jun N-terminal kinases (JNKs).

Brand:  Invitrogen™ PV4564

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15448733

  • £6900.00 / 1mg
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MAPK10 is involved in neuronal apoptosis and phosphorylates MAP2K4.

Specifications

Specifications

NP_002744
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MAPK10 (JNK3)
Human
GST Tag
Frozen
Nuclear Receptor Assay
76.6kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
MAP Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers