missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP4K6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49220-25ul
This item is not returnable.
View return policy
Description
MAP4K6 Polyclonal antibody specifically detects MAP4K6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MAP4K6 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| B55, EC 2.7.11, GCK family kinase MiNK, hMINK, hMINKbeta, MAP4K6EC 2.7.11.1, MAPK/ERK kinase kinase kinase 6, MEK kinase kinase 6, MGC21111, MINKMEKKK 6, Misshapen/NIK-related kinase, misshapen-like kinase 1, misshapen-like kinase 1 (zebrafish), Mitogen-activated protein kinase kinase kinase kinase 6, serine/threonine protein kinase, YSK2, ZC3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ARPRSNSAWQIYLQRRAERGTPKPPGPPAQPPGPPNASSNPDLRRSDPGWERSDSVLPASHGHL | |
| 25 μL | |
| MAP Kinase Signaling, Protein Kinase | |
| 50488 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction