missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
£270.00 - £432.00
Specifications
| Antigen | MAP2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646018
|
Novus Biologicals
NBP3-05552-100ul |
100 μg |
£432.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689747
|
Novus Biologicals
NBP3-05552-50ul |
50 μg |
£270.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP2 Polyclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MAP2 | |
| Western Blot 1:2000, Immunohistochemistry 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Goat | |
| Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers | |
| 50% PBS, 50% glycerol | |
| 4133 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title