missing translation for 'onlineSavingsMsg'
Learn More

MAP2 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP3-05552-50ul

 View more versions of this product

Product Code. 18689747

  • £270.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MAP2 Polyclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MAP2
Polyclonal
Unconjugated
50% PBS, 50% glycerol
Goat
Immunogen affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:1000-1:2000
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
MAP2
50 μg
Cell Biology, Cellular Markers, MAP Kinase Signaling, Neuroscience, Stem Cell Markers
4133
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.