missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | MAP1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622836
|
Novus Biologicals
NBP2-98818-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18656795
|
Novus Biologicals
NBP2-98818-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP1 Polyclonal antibody specifically detects MAP1 in Human samples. It is validated for Western BlotSpecifications
| MAP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, MAP Kinase Signaling | |
| 0.2 um filtered solution in PBS | |
| 64112 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MAP1, MAP-1PNMA4Paraneoplastic antigen Ma4, modulator of apoptosis 1, paraneoplastic antigen like 4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human MAP1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title