missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98818-50ul
This item is not returnable.
View return policy
Description
MAP1 Polyclonal antibody specifically detects MAP1 in Human samples. It is validated for Western Blot
Specifications
| MAP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MAP1, MAP-1PNMA4Paraneoplastic antigen Ma4, modulator of apoptosis 1, paraneoplastic antigen like 4 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human MAP1. | |
| 50 μg | |
| Apoptosis, MAP Kinase Signaling | |
| 64112 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction