missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Manual Ergonomic Crimper

Product Code. 12888585 Shop All Macherey-Nagel Products
Change view
Click to view available options
Size:
11 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12888585 11 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12888585 Supplier Macherey-Nagel™ Supplier No. 735211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

No. per Pack 1 Each
Type Manual Ergonomic Crimper
For Use With (Equipment) For 11mm Aluminum Crimp Caps
Size 11 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.