Learn More
Macherey-Nagel™ Manual Crimper
Product Code. 10419561 Shop All Macherey-Nagel Products
Change view
Click to view available options
For Use With (Equipment):
11 mm Aluminum Crimp Caps
13 mm Aluminum Crimp Caps
20 mm Aluminum Crimp Caps
8 mm Aluminum Caps
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10419561 8 mm Aluminum Caps Each
10003061 13 mm Aluminum Crimp Caps Each
10216911 20 mm Aluminum Crimp Caps Each
10246811 11 mm Aluminum Crimp Caps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10419561 Supplier Macherey-Nagel™ Supplier No. 735126