missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD™ Mannitol Salt Agar Plates
Description
Mannitol Salt Agar is a formulation devised by Chapman for the differentiation of coagulase positive staphylococci (e.g., Staphylococcus aureus) from coagulase negative staphylococci.
- Used for isolating staphylococci from clinical specimens, from cosmetics, and in microbial limit tests.
- Contains peptones and beef extract, which supply essential nutrients.
- The 7.5% concentration of sodium chloride results in the partial or complete inhibition of bacterial organisms other than staphylococci.
- Mannitol fermentation, as indicated by a change in the phenol red indicator, aids in the differentiation of staphylococcal species.
- Positive staphylococci produce yellow colonies and a yellow medium; negative staphylococci produce red colonies and no colour change of the indicator.
Supplied in ready to use Stacker™ plates
- Prepared plated media are ready for immediate use
- Microbiologically controlled for uncompromised isolation of microorganisms
- Specialized lid and bottom allow plates to interlock and stack without slipping
- Specially engineered design easily separates without agitating the sample
Specifications
Specifications
| Certifications/Compliance | EP/USP |
| Content And Storage | Store in dark in original sleeve in 2°-8° C |
| Detects | Staphylococci |
| Format | Prepared culture plates |
| Product Type | Agar Plate |
| Target Organism Class | S. aureus |
| Formulation | Per 1 L prepared: Beef Extract 1 g, Pancreatic Digest of Casein 5 g, Peptic Digest of Animal Tissue 5 g, Sodium Chloride 75 g, D-Mannitol 10 g, Phenol Red 0.025 g, Agar 15 g |
| For Use With (Application) | Selective isolation of staphylococci and the detection of Staphylococcus aureus |
| Incubation Conditions | 35 ± 2° C in an aerobic atmosphere |
| Incubation Time | 48-72 hours |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?