missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mannitol Salt Agar (EP, USP, JP, BP)

Product Code. 11904142
This item is not returnable. View return policy

Product Code. 11904142

Brand: Thermo Scientific™ PO1169A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens with this ready-to-use medium.

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other samples with ready-to-use Thermo Scientific™ Oxoid™ Mannitol Salt Agar (EP, USP, JP, BP). Due to high concentrations of salt in the medium, most unwanted bacteria are inhibited with the exception of some halophilic marine organisms. Presumptive coagulase-positive staphylococci produce colonies surrounded by bright yellow zones while non-pathogenic staphylococci produce colonies with reddish purple zones. The medium is tested in accordance with International Pharmacopoeia.1,2,3,4

Staphylococcus aureus is a coagulase-positive, catalase-positive, Gram-positive cocci that occurs singly or in pairs, tetrads, short chains, and grape-like clusters. S. aureus can produce a variety of syndromes with clinical manifestations, including skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis. Use Mannitol Salt Agar (EP, USP, JP, BP) for clear and easy differentiation of Staphylococcus aureus and Staphylococcus epidermis from food samples while most other bacteria are inhibited by the high salt concentration.

  • Highly selective due to high concentrations of salt in the medium.
  • Easy to differentiate: Staphylococcus aureus forms yellow colonies with yellow halo while Staphylococcus epidermis forms pink colonies with pink medium.
  • Conforms to formulations detailed in International Pharmacopoeia Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Red, clear
Content And Storage 2°C to 10°C
Description Mannitol Salt Agar (EP, USP, JP, BP)
Format 90mm Plate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.