missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mannitol Salt Agar

Product Code. 10473663
Change view
Click to view available options
Format:
90mm Monoplate
Color:
Antique pink, transparent
Red, clear
Manufacturing Origin:
Germany
United Kingdom
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Color Manufacturing Origin Unit Size
10473663 90mm Monoplate Antique pink, transparent Germany Pack of 10
12929420 90mm Monoplate Red, clear United Kingdom Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10473663 Supplier Thermo Scientific™ Supplier No. PO5027A
  • ONLINE EXCLUSIVE PRICE
    £13.80
    £13.54 / Pack of 10
    Save £0.26
    1% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens while most other bacteria are inhibited.

Presumptively detect, isolate and enumerate pathogenic staphylococci in milk, food, fish and other specimens while most other bacteria are inhibited, except a few halophilic species, with Thermo Scientific™ Mannitol Salt Agar. Look for colonies surrounded by bright yellow zones to recognize coagulase-positive staphylococci; non-pathogenic staphylococci produce colonies with reddish purple zones.

Clearly and easily differentiate Staphylococcus aureus and Staphylococcus epidermidis while most other bacteria is inhibited by the high salt concentration.

Staphylococcus aureus is a coagulase-positive, catalase-positive, gram-positive cocci that occurs singly or in pairs, tetrads, short chains, and grape-like clusters. It is commonly carried on the skin and in the nose of healthy individuals but is also a major cause of infection in both healthcare and community settings worldwide. S. aureus can produce a variety of syndromes with clinical manifestations, including
skin and soft tissue infections, empyema, bloodstream infections, pneumonia, osteomyelitis, septic arthritis, endocarditis, sepsis, and meningitis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Antique pink, transparent
Content And Storage 2° to 12°C, in the dark
Description Mannitol Salt Agar
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow wrapped - single layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.