missing translation for 'onlineSavingsMsg'
Learn More

Maltitol, TRC

Product Code. 30138405
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138405 25 g 25g
30136207 50 g 50g
30005365 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138405 Supplier Toronto Research Chemicals Supplier No. TRCM13501025G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-88-6
Molecular Formula C12 H24 O11
Molecular Weight (g/mol) 344.31
Synonym Glucitol, 4-O-α-D-glucopyranosyl-, D- (8CI), Maltitol (6CI,7CI), 4-O-α-D-Glucopyranosyl-D-glucitol, Amalty, Amalty MR, Amalty MR 100, Amalty MR 20, Amalty MR 50, Amalty P, Amalty Syrup, Cerestar 16303, D-Maltitol, Lesys, Lycasin HBC, Mabit, Mabit 50M, Malbit CH, Malbit CH 16385, Malbit CR, Malti MR, Maltidex, Maltidex 100, Maltidex CH 16385, Maltidex H 16330, Maltidex M 16313, Maltilite 5575, Maltilite 7575, Maltilite P 200, Maltilol, Maltisorb, Maltisorb 75/77, Maltisorb P 200, Maltisorb P 35, Maltisorb P 90, Maltisweet, Maltisweet 3145, Maltit, Maltitol G 3, Sweet G 2, SweetPearl, SweetPearl P 200, SweetPearl P 35
IUPAC Name (2S,3R,4R,5R)-4-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexane-1,2,3,5,6-pentol
SMILES OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)CO

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H24O11/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12/h4-21H,1-3H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+/m0/s1
Formula Weight 344.1319
Percent Purity >95%
Chemical Name or Material Maltitol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.