missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Malt Extract Agar Contact Plate

Product Code. 10026782
This item is not returnable. View return policy
Product Code. 10026782 Supplier Thermo Scientific™ Supplier No. PO5079C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect, isolate and enumerate yeasts and molds with this formulation in contact plate format.

Detect, isolate and enumerate yeasts and molds with ready-to-use Thermo Scientific™ Oxoid™ Malt Extract Agar Contact Plate. Mycological peptone in the medium gives a luxuriant growth of yeasts and molds with typical morphology and pigmentation. Due to acidic nature of the medium, it inhibits the growth of bacteria resulting in improved isolation of yeasts and molds from materials heavily infected with bacteria.

Perform effortless surface sampling with easy-to-handle Malt Extract Agar Contact Plate.

  • Supports luxuriant growth of yeasts and molds while inhibiting the growth of bacteria
  • Easy-to-handle dome-shaped bottom format for effortless surface sampling.
  • Consistent performance: smooth, even agar surface ensured through specialized manufacturing process
  • Easy-to-stack providing more stability during handling and transport
  • Ready-to-use convenience of prepared media.

Prevent agar from falling out of the dish with Thermo Scientific Contact Plates, which feature a dome-shaped bottom. GripFit™ technology makes plates easy-to-handle and ensures the lid remains in place during handling, while being easily removed during environmental monitoring. An inner layer on the base of the plate is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits. Easy stacking provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out.

Our contact plates are manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate. Surface sampling is achieved by gently rolling the domed surface of the agar onto the area to be tested. If any microorganisms that can cause disease or infection are present, they will grow to form easily visible colonies on the surface of the agar. This offers a reliable and effective way of determining the microbial cleanliness of surface areas and help control invisible dangers associated with microbial contamination. Clearly visible growth is available within three days for bacteria and within five days for fungi. 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Ocher yellow, transparent
Content And Storage 2 to 25°C
Description Malt Extract Agar Contact Plate
Format Contact Plate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 13.5g ± 5 %
Packaging Type Boxes with 2 x 10 plates wrapped in film
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.