missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Malachite Green isothiocyanate

Product Code. 11539106
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11539106 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11539106 Supplier Invitrogen™ Supplier No. M689

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

malachite green isothiocyanate

Malachite green is a nonfluorescent photosensitizer that absorbs at long wavelengths (∼630 nm). Its photosensitizing action can be targeted to particular cellular sites by using the amine-reactive malachite green isothiocyanate to conjugate this label to antibodies or other biomolecules.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type Malachite Green Isothiocyanate
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Isothiocyanate
Label or Dye Malchite Green
Label Type Other Labels or Dyes
Color Green

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.