missing translation for 'onlineSavingsMsg'
Learn More

MAL2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£161.00 - £378.00

Specifications

Antigen MAL2
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18331295
View Documents
Novus Biologicals
NBP3-10113-25UL
25 μg
£161.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18330876
View Documents
Novus Biologicals
NBP3-10113-100UL
100 μg
£378.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MAL2 Polyclonal specifically detects MAL2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MAL2
Western Blot
Unconjugated
Rabbit
Mouse
MAL proteolipid protein 2, mal, T-cell differentiation protein 2 (gene/pseudogene), MAL2 proteolipid protein, protein MAL2
The immunogen is a synthetic peptide directed towards the N terminal region of Mouse MAL2 (NP_849251.1). Peptide sequence MSAGGAVPPPPNPAVSFPAPRVTLPAGPDILRTYSGAFVCLEIVLGGLVW
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
114569
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.