missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Magnetic Antibody Labeling Kit

Product Code. 18067094 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
200µg
This item is not returnable. View return policy

Product Code. 18067094

Brand: Novus Biologicals™ 13000015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

The Magnetic Conjugation Kit allows one-step conjugation of antibodies, proteins and peptides (or any other biomolecule with an amine group) to specially treated magnetic particles without the need for extensive optimization of the conjugation reaction.

Kit Contents:

  • 1 Midi vial of 500 nm (0.5 μm) Magnetic particles
  • 1 bottle of Mag Storage Buffer
  • 1 vial/bottle of Mag Wash Buffer
  • 1 vial/bottle of Mag Antibody Diluent
  • 1 vial/bottle of Mag Quencher
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Antibody Labeling Kit
Content And Storage 4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.