missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitride, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog Number 11344927
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
12057-71-5
MFCD00016205
This item is not returnable. View return policy
12057-71-5
MFCD00016205

The use of magnesium nitride as a convenient source of ammonia allows a direct transformation of esters to primary amides. Methyl, ethyl, isopropyl, and tert-butyl esters are converted to the corresponding carboxamides in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 800°C (decomposition)
Physical Form Powder
Quantity 10 g
UN Number UN3178
Solubility Information Reacts with water.
Formula Weight 100.95
Percent Purity 99.6%
Sensitivity Moisture sensitive
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium nitride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'