CAS RN 12057-71-5
IUPAC Name:
diazenediide; trimagnesium-1,3-bis(ylium)
Synonyms:
magnesium nitride
Molecular Weight (g/mol):
100.93
Molecular Formula:
Mg3N2
InChi Key:
VZPQZBKNAUHPOS-UHFFFAOYSA-N
SMILES:
[N-]=[N-].[Mg+][Mg][Mg+]
1
–
1
of
1
results
1
Magnesium nitride, 99.6% (metals basis)
CAS: 12057-71-5 MDL Number: MFCD00016205