missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitride, 99.6% (metals basis)

Product Code. p-7051776
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11344927

Brand: Thermo Scientific Alfa Aesar 041946.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12057-71-5

The use of magnesium nitride as a convenient source of ammonia allows a direct transformation of esters to primary amides. Methyl, ethyl, isopropyl, and tert-butyl esters are converted to the corresponding carboxamides in good yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12057-71-5
MDL Number MFCD00016205

Specifications

Melting Point 800°C (decomposition)
Physical Form Powder
Quantity 10 g
UN Number UN3178
Solubility Information Reacts with water.
Formula Weight 100.95
Percent Purity 99.6%
Sensitivity Moisture sensitive
Density 2.71 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium nitride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.