missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate GFAA Matrix Modifier: 1% Mg(NO3)2 in 2% HNO3, VHG™

Product Code. 30003039
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003039 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003039 Supplier VHG Labs Supplier No. VHGMMGN1P100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Magnesium Nitrate GFAA Matrix Modifier: 1% Mg(NO3)2 in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Nitrate
CAS 10377-60-3
Methods AA/GFAA
Analyte or Component Names Magnesium Nitrate
Shipping Condition Room Temperature
Molecular Formula Mg(NO3)2
Concentration 1%
Synonym Mg(NO3)2
SMILES [Mg+2].[O-][N+](=O)[O-].[O-][N+](=O)[O-]
IUPAC Name Magnesium Nitrate
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 148.31
Quantity 100 mL
Formula Weight 148.31 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.