missing translation for 'onlineSavingsMsg'
Learn More

Magnesium molybdenum oxide, 99.9% (metals basis)

Product Code. 11328148
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328148 50 g 50g
11338148 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328148 Supplier Thermo Scientific Alfa Aesar Supplier No. 039376.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13767-03-8 | MgMoO4 | 184.251 g/mol

Magnesium molybdenum oxide is used as an intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13767-03-8
Molecular Formula MgMoO4
Molecular Weight (g/mol) 184.251
MDL Number MFCD00016195
InChI Key MODMKKOKHKJFHJ-UHFFFAOYSA-N
Synonym magnesium molybdate, magnesium molybdenum oxide, unii-ou5m6mzi9r, ou5m6mzi9r, magnesium dioxido dioxo molybdenum, mg.moo4, magnesium-molybdate, magnesium 2+ molybdate, magnesium 2+ ion molybdate
PubChem CID 3084170
IUPAC Name magnesium;dioxido(dioxo)molybdenum
SMILES [O-][Mo](=O)(=O)[O-].[Mg+2]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 184.25
Percent Purity 99.9%
Odor Odorless
Density 2.208 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium molybdenum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.