accessibility menu, dialog, popup

UserName

MgOMoOOO

CAS RN 13767-03-8

IUPAC Name: magnesium(2+) dioxomolybdenumbis(olate)
Synonyms: magnesium(2+) molybdate
Molecular Weight (g/mol): 184.25
Molecular Formula: MgMoO4
InChi Key: MODMKKOKHKJFHJ-UHFFFAOYSA-N
SMILES: [Mg++].[O-][Mo]([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results