missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAD2L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | MAD2L1 |
|---|---|
| Concentration | 0.21 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16866663
|
Proteintech
10337-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16856663
|
Proteintech
10337-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAD2L1 is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. MAD2L1 is related to the MAD2L2 gene located on chromosome 1. A MAD2 pseudogene has been mapped to chromosome 14.Specifications
| MAD2L1 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| Q13257, Q9Z1B5 | |
| 297176, 4085, 56150 | |
| MAD2L1 Fusion Protein Ag0307 | |
| Primary | |
| -20°C | |
| MAD2L1 |
| 0.21 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| HSMAD2, MAD2, MAD2 like protein 1, MAD2L1 | |
| Mad2l1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit