missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar No.3 (Contact)

Product Code. 10321893
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10321893 10 Plates Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321893 Supplier Thermo Scientific™ Supplier No. PO5053C
  • ONLINE EXCLUSIVE PRICE
    £9.20
    £8.99 / Pack of 10
    Save £0.21
    2% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Optimize microbial isolation with this agar, designed for targeted detection of Gram-negative bacteria.

Optimize microbial isolation with MacConkey Agar No.3 (Contact), designed for targeted detection of Gram-negative bacteria.

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Content And Storage 2°C to 12°C
Format 55 mm
Manufacturing Origin Germany
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.