missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar (EP, USP, JP, BP)

Product Code. 13215349
This item is not returnable. View return policy
Product Code. 13215349 Supplier Thermo Scientific™ Supplier No. PO5002A
  • ONLINE EXCLUSIVE PRICE
    £11.43
    £8.59 / Pack of 10
    Save £2.84
    24% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with this ready-to-use medium.

Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with ready-to-use Thermo Scientific™ Oxoid™ MacConkey Agar (EP, USP, JP, BP). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water. The medium conforms to the formulations detailed in international pharmacopoeia3,4,5,6.

Use MacConkey Agar (EP, USP, JP, BP) for the cultivation of pathogens which may be present in a variety of specimens .

  • Easy-to-read: easy to differentiate Escherichia coli (red non-mucoid colonies) from Staphyloccoccus spp. (pale pink opaque colonies) and Pseudomonas aeruginosa (green-brown) colonies.
  • Recommended by World Health Organizaton1, the Department of Health 2
  • Conforms to formulations detailed in International Pharmacopoeia3,4,5,6,
  • Ready-to-use convenience of prepared media.

Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Dark Red, clear
Content And Storage 2°C to 10°C
Description MacConkey Agar (EP, USP, JP, BP)
Format 90mm Plate
Manufacturing Origin United Kingdom
Product Type Agar Plate
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.