missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MabSelect Xtra™ Chromatography Media
Provides high purity capture and high dynamic binding capacity
£2391.00 - £5090.00
Specifications
For Use With (Application) | For Develop to Meet The Demands of Increasing Levels of Expression in Monoclonal Antibody Feedstocks |
---|---|
Storage Requirements | 2°C to 8°C |
Binding Capacity | 40 mg Human lgG/mL (at 2.4 min. residence time) |
Product Type | MabSelect Xtra Media |
Flow Rate | <300 cm/Hoµr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11389981
|
Cytiva
17-5269-07 |
25 mL |
£2391.00
25mL |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11379981
|
Cytiva
17-5269-02 |
200 mL |
£5090.00
200mL |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
- High ligand selectivity and matrix hydrophilicity
- Improved process economics through reduced raw materials costs and/or reduced number of cycles
- Slightly lower pressure/flow properties compared to MabSelect™ SuRe™ and MabSelect
Specifications
For Develop to Meet The Demands of Increasing Levels of Expression in Monoclonal Antibody Feedstocks | |
40 mg Human lgG/mL (at 2.4 min. residence time) | |
<300 cm/Hoµr | |
7.87 in. | |
<200 kPa | |
2 to 12 | |
Recombinant Protein A (E. coli) |
2°C to 8°C | |
MabSelect Xtra Media | |
75 μm | |
20 cm | |
Highly Cross-Linked Agarose | |
20% Ethanol |