missing translation for 'onlineSavingsMsg'
Learn More

Cytiva MabSelect Xtra™ Chromatography Media

Product Code. 11389981 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
200 mL
Unit Size:
200mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389981 25 mL 25mL
11379981 200 mL 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389981 Supplier Cytiva Supplier No. 17526907

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides high purity capture and high dynamic binding capacity

  • High ligand selectivity and matrix hydrophilicity
  • Improved process economics through reduced raw materials costs and/or reduced number of cycles
  • Slightly lower pressure/flow properties compared to MabSelect™ SuRe™ and MabSelect

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
For Use With (Application) For Develop to Meet The Demands of Increasing Levels of Expression in Monoclonal Antibody Feedstocks
Storage Requirements 2°C to 8°C
Binding Capacity 40 mg Human lgG/mL (at 2.4 min. residence time)
Product Type MabSelect Xtra Media
Flow Rate <300 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Max. Pressure <200 kPa
Matrix Highly Cross-Linked Agarose
pH Range 2 to 12
Storage Buffer 20% Ethanol
Ligand Type Recombinant Protein A (E. coli)
Show More Show Less
compliance-icons
Product Identifier
  • MabSelect Xtra
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.