missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ M13 phage coat protein Monoclonal Antibody (A5B3), HRP

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA536125
325.50 GBP valid until 2025-12-19
BEST PRICE on Promo! Use promo code "24110" to get your promotional price.
This item is not returnable.
View return policy
Description
M13 phage coat protein Monoclonal Antibody for ELISA
M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.
Specifications
| M13 phage coat protein | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 40% glycerol, 0.1% BSA and no preservative; pH 7.4 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA | |
| A5B3 | |
| HRP | |
| Attachment protein G3P; G3P; Gene 3 protein; III; Minor coat protein | |
| Synthetic peptide within bacteriophage M13 aa 1-50. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction