missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-Ras/R-Ras3 Antibody (18E11), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | M-Ras/R-Ras3 |
|---|---|
| Clone | 18E11 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032727
|
Novus Biologicals
NBP3-26129-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30036403
|
Novus Biologicals
NBP3-26129-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
M-Ras/R-Ras3 Monoclonal antibody specifically detects M-Ras/R-Ras3 in Human samples. It is validated for Western Blot, ELISASpecifications
| M-Ras/R-Ras3 | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
| A synthesized peptide derived from Human M-Ras/R-Ras3 [UniProt O14807] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 18E11 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 22808 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title