missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M-Ras/R-Ras3 Antibody (18E11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26129-100ul
This item is not returnable.
View return policy
Description
M-Ras/R-Ras3 Monoclonal antibody specifically detects M-Ras/R-Ras3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| M-Ras/R-Ras3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 18E11 | |
| Western Blot 1:500-1:2000, ELISA | |
| FLJ42964, muscle and microspikes RAS, muscle RAS oncogene homolog, ras-related protein M-Ras, Ras-related protein R-Ras3, R-RAS3, RRAS3M-RAs | |
| A synthesized peptide derived from Human M-Ras/R-Ras3 [UniProt O14807] | |
| 100 μL | |
| Signal Transduction | |
| 22808 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction