missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Rabbit anti-Human, Clone: 9F0W6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Lysyl Oxidase Homolog 2/LOXL2 |
|---|---|
| Clone | 9F0W6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18337301
|
Bio-Techne
NBP3-16466-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18309486
|
Novus Biologicals
NBP3-16466-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Western BlotSpecifications
| Lysyl Oxidase Homolog 2/LOXL2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4017 | |
| IgG | |
| Affinity purified |
| 9F0W6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 | |
| A synthetic peptide corresponding to a sequence within amino acids 675-774 of human Lysyl Oxidase Homolog 2/LOXL2 (Q9Y4K0). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title