missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysyl Oxidase Homolog 2/LOXL2 Rabbit anti-Human, Clone: 9F0W6, Novus Biologicals™
Description
Lysyl Oxidase Homolog 2/LOXL2 Monoclonal antibody specifically detects Lysyl Oxidase Homolog 2/LOXL2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Lysyl Oxidase Homolog 2/LOXL2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 9F0W6 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 675-774 of human Lysyl Oxidase Homolog 2/LOXL2 (Q9Y4K0). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?