Learn More
Invitrogen™ LysoLight™ Antibody Labeling Kits

Description
LysoLight Antibody Labeling Kits provide a powerful tool for visualizing the lysosomal localization and catabolic degradation of antibodies and proteins. These kits utilize a unique fluorophore that is only active upon enzymatic cleavage by the lysosomal specific enzyme, Cathepsin B. Outside of the lysosome, LysoLight dyes have little to no observable background fluorescence, and only upon localization to the protease rich environment of the lysosome, does LysoLight become activated and brightly fluoresce bright green or deep red in color.
Specifications
Specifications
| Color | Deep Red |
| Description | LysoLight™ Deep Red Amine Reactive Dye |
| Detection Method | Fluorescence |
| For Use With (Application) | Antibody and Protein Labeling |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope, High Content Analysis Instrument |
| Product Type | Amine Reactive Dye |
| Green Features | Enhanced stability |
| Product Line | LysoLight |
| Quantity | 3 x 100 μg |
| Solubility | DMSO (Dimethylsulfoxide) |
| Show More |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.