missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Lysing Matrix E Barcoded, 96-Well Plate

Product Code. 15749200 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 Rack
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15749200

Brand: MP Biomedicals 116984001B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.

  • Rack includes 1.2 mL cluster-strip tube and cap assemblies.
  • Each tube is filled with 1.4 mm ceramic spheres, 0.1 mm silica spheres, 1 x 4 mm glass bead.
  • Individual tubes are 2-D barcoded.
  • Will not bind nucleic acids.
  • Key applications: Nucleic Acid Isolation, Nucleic Acid Extraction, Cell culture, Proteomics, Purification and Detection, Protein Purification, DNA Purification, RNA Purification, Plant Tissue Processing, Tissue Analysis, Soil Analysis, Bacterial Culture, Yeast Culture, Fungal Tissue Analysis, Microbiome Studies, Metabolite Extraction, Environmental Analysis, Lysis of Organisms in Feces, Fecal Metagenomics, Wastewater Analysis, Water Analysis, Sludge Analysis, Tumor Homogenization .
  • Research Use Only (RUO).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Rack
Includes Matrix E: 1.4 ceramic spheres, 0.1 mm silica spheres, and 1 (one) 4 mm glass bead. Cluster-strip tube, polypropylene. Cluster-strip cap, silicone, Deep rack, polypropylene
Material Ceramic, silica, glass, polypropylene
Content And Storage Store at room temperature (15-30 °C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.