Learn More
Thermo Scientific™ Lysed GC Selective Agar
Description
Lysed GC Selective Agar (PB1205A) is a qualitative diagnostic media for the isolation of pathogenic Neisseria species (Neisseria gonorrhoeae, Neisseria meningitidis). Media can be used with a variety of clinical samples; the main sample types are rectal, pharyngeal, cervical and urethral swabs.
Lysed GC Selective Agar (PB1205A) is a diagnostic tool to aid clinicians in determining potential treatment options for patients suspected of having bacterial infections. The devices are for professional use only, are not automated and nor are they for companion diagnostics.
Use Lysed GC Selective Agar for the isolation of Neisseria species from clinical specimens.
- Supports the growth of Neisseria species with chocolated blood providing the nutritional requirements.
- Improved isolation of Neisseria gonorrhoeae and Neisseria meningitidis due to selective agents reducing competing respiratory flora.
- Ready-to-use convenience of prepared media.
Specifications
Specifications
| Content And Storage | 2° to 10°C |
| Description | Lysed GC Selective Agar |
| For Use With (Application) | Clinical samples |
| Quantity | 10 plates |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.